Skip to Content
Merck
All Photos(3)

Key Documents

V900377

Sigma-Aldrich

Putrescine dihydrochloride

Vetec, reagent grade, ≥98%

Synonym(s):

1,4-Butanediamine dihydrochloride, 1,4-Diaminobutane dihydrochloride, Tetramethylenediamine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)4NH2 · 2HCl
CAS Number:
Molecular Weight:
161.07
Beilstein:
3906680
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

≥98%

mp

280 °C (dec.) (lit.)

SMILES string

Cl[H].Cl[H].NCCCCN

InChI

1S/C4H12N2.2ClH/c5-3-1-2-4-6;;/h1-6H2;2*1H

InChI key

XXWCODXIQWIHQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Binds to the polyamine modulatory site of the NMDA receptor and potentiates NMDA-induced currents; precursor of spermidine.
Binds to the polyamine modulatory site of the NMDA receptor; potentiates NMDA-induced currents.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Tao et al.
Human reproduction (Oxford, England), 30(8), 1867-1875 (2015-06-18)
Does peri-ovulatory putrescine supplementation of older mice improve oocyte quality and reduce the incidence of embryo resorption? Peri-ovulatory putrescine supplementation in older mice improved oocyte quality, as indicated by increased blastocyst cell numbers and reduced the incidence of embryo resorption.
Elvira S Plakidi et al.
Foods (Basel, Switzerland), 9(5) (2020-05-14)
Monitoring of biogenic amines in food is important for quality control, in terms of freshness evaluation and even more for food safety. A novel and cost-effective method was developed and validated for the determination of the main biogenic amines: histamine
Injae Hwang et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(6), 2397-2411 (2015-02-26)
Firmicutes and Bacteroidetes, 2 major phyla of gut microbiota, are involved in lipid and bile acid metabolism to maintain systemic energy homeostasis in host. Recently, accumulating evidence has suggested that dietary changes promptly induce the alteration of abundance of both
Quan Findlay et al.
American journal of physiology. Gastrointestinal and liver physiology, 307(7), G741-G748 (2014-08-16)
Gut motility disorders can result from an absent, damaged, or dysfunctional enteric nervous system (ENS). Cell therapy is an exciting prospect to treat these enteric neuropathies and restore gut motility. Previous studies have examined a variety of sources of stem/progenitor
Suk Ho Bhang et al.
Biomaterials, 55, 33-43 (2015-05-03)
At high concentrations, manganese (Mn) promotes cellular neurodevelopment but causes toxicity. Here, we report that Mn ion at high concentrations can be delivered to pheochromocytoma 12 (PC12) cells using gold nanoparticles (AuNPs) to enhance cellular neurodevelopment without toxicity. Mn(2+) release

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service