Skip to Content
Merck
All Photos(1)

Key Documents

V900276

Sigma-Aldrich

Cobalt(II) sulfate heptahydrate

Vetec, reagent grade, 99%

Synonym(s):

Cobaltous sulfate heptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoSO4 · 7H2O
CAS Number:
Molecular Weight:
281.10
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

99%

form

powder

pH

4 (20 °C, 100 g/L)

density

2.03 g/mL at 25 °C (lit.)

SMILES string

O.[Co++].[O-]S([O-])(=O)=O

InChI

1S/Co.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+2;;/p-2

InChI key

BGORGFZEVHFAQU-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali M Elshafei et al.
Journal of basic microbiology, 45(5), 335-343 (2005-09-28)
Cell-free extracts of nitrate-grown Penicillium politans NRC-510 catalyzes the hydrolytic deamination of cytidine to uridine. Uridine was chromatographically identified in cell-free extracts. The enzyme exhibited optimum pH and temperature activities at 6.5 and 80 degrees C respectively. Thermal stability experiments
Y J Huang et al.
Chemosphere, 40(4), 347-349 (2000-02-09)
A rapid and complete destruction of organics in the supercritical water oxidation (SCWO) of the Co-exchanged resin was found experimentally. Due to an extremely low solubility of CoSO4 salt formed and separated effectively in the SCWO process, a minimal release
Hong Wang et al.
ACS nano, 7(1), 614-626 (2012-12-12)
Electronic and optical properties of single-walled carbon nanotubes (SWCNTs) correlate with their chiral structures. Many applications need chirally pure SWCNTs that current synthesis methods cannot produce. Here, we show a sulfate-promoted CoSO(4)/SiO(2) catalyst, which selectively grows large-diameter (9,8) nanotubes at
P J Duerksen-Hughes et al.
Environmental health perspectives, 107(10), 805-812 (1999-09-30)
In vivo carcinogenicity testing is an expensive and time-consuming process, and as a result, only a relatively small fraction of new and existing chemicals has been tested in this manner. Therefore, the development and validation of alternative approaches is desirable.
Lei Hou et al.
Chemical communications (Cambridge, England), 46(34), 6311-6313 (2010-08-03)
Solvothermal reaction of CoSO(4) with 2,6-di-p-carboxyphenyl-4,4'-bipyridine affords a novel, octacobalt cluster based, (3,12)-connected porous framework, which exhibits gas sorption and spin-glassy magnetic behaviour.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service