Skip to Content
Merck
All Photos(1)

Documents

V800305

Sigma-Aldrich

Phosphorus pentoxide

LR, ≥95%

Synonym(s):

Phosphoric anhydride, Phosphorus(V) oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
P2O5
CAS Number:
Molecular Weight:
141.94
EC Number:
UNSPSC Code:
12352303

grade

LR

vapor density

4.9 (vs air)

vapor pressure

1 mmHg ( 384 °C)
10 mmHg ( 238 °C)

product line

Vetec

Assay

≥95%

form

powder

pH

1.5 (20 °C, 10 g/L)

mp

340 °C (lit.)

density

2.3 g/mL at 25 °C (lit.)

SMILES string

O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3

InChI

1S/O10P4/c1-11-5-12(2)8-13(3,6-11)10-14(4,7-11)9-12

InChI key

YWEUIGNSBFLMFL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Devalina Ray et al.
Carbohydrate research, 346(14), 2145-2148 (2011-09-06)
A convenient, mild and environment-friendly dehydration reaction of fructose in ionic liquid using phosphorous pentoxide (P(2)O(5)) has been investigated. The acidic nature of P(2)O(5) along with its hygroscopic properties has been successfully utilized to afford 81.2% yield of 5-hydroxymethylfurfural (5-HMF)
Radek Pohl et al.
Organic & biomolecular chemistry, 13(16), 4693-4705 (2015-03-24)
Systematic structure-activity studies on a diverse family of nucleoside phosphonic acids has led to the development of potent antiviral drugs such as HPMPC (CidofovirTM), PMEA (AdefovirTM), and PMPA (TenofovirTM), which are used in the treatment of CMV-induced retinitis, hepatitis B
Jae-Hyung Ahn et al.
Microbial ecology, 68(2), 271-283 (2014-04-01)
The effects of water-saving irrigation on emissions of greenhouse gases and soil prokaryotic communities were investigated in an experimental rice field. The water layer was kept at 1-2 cm in the water-saving (WS) irrigation treatment and at 6 cm in
J Ma et al.
Colloids and surfaces. B, Biointerfaces, 81(1), 87-95 (2010-07-30)
Mesoporous 58SiO(2)-(38-x)CaO-xMgO-4P(2)O(5) glasses (where x=0, 5, 10 and 20 mol%) have been prepared by the sol-gel synthesis route. The effects of the substitution of MgO for CaO on glass degradation and bioactivity were studied in tris-(hydroxymethyl)-aminomethane and hydrochloric acid buffer
Ana García et al.
Acta biomaterialia, 7(3), 1265-1273 (2010-10-19)
Herein we report for the first time the synthesis of three-dimensional scaffolds in the binary system SiO2-P2O5 exhibiting different scales of porosity: (i) highly ordered mesopores with diameters of ca. 4 nm; (ii) macropores with diameters in the 30-80 μm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service