Skip to Content
Merck
All Photos(2)

Documents

V800148

Sigma-Aldrich

di-Sodium hydrogen phosphate dihydrate

LR, ≥98%

Synonym(s):

sec-Sodium phosphate, Disodium hydrogen phosphate dihydrate, Disodium phosphate, Sodium phosphate dibasic dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4 · 2H2O
CAS Number:
Molecular Weight:
177.99
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

LR

product line

Vetec

Assay

≥98%

form

powder

pH

8.4-9.6 (5% in water)

SMILES string

O.O.[Na+].[Na+].OP([O-])([O-])=O

InChI

1S/2Na.H3O4P.2H2O/c;;1-5(2,3)4;;/h;;(H3,1,2,3,4);2*1H2/q2*+1;;;/p-2

InChI key

KDQPSPMLNJTZAL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrizia Iavicoli et al.
Journal of chromatography. A, 1422, 260-269 (2015-10-27)
Asymmetric Flow Field-Flow Fractionation (AF4) combined with multidetector analysis form a promising technique in the field of nanoparticle characterization. This system is able to measure the dimensions and physicochemical properties of nanoparticles with unprecedented accuracy and precision. Here, for the
Christopher Polonyi et al.
Dalton transactions (Cambridge, England : 2003), 43(31), 11941-11949 (2014-06-27)
The ability of the anticancer active drug cisplatin to exert biological activity through interference with nucleic acid function is well documented. Since kinetics play a key role in determining product distributions in these systems, methods for accurate documentation of reactivity
Brecht Gorissen et al.
Analytical and bioanalytical chemistry, 407(15), 4447-4457 (2015-01-31)
The most important source of Salmonella spp. infection in humans is by the consumption of contaminated poultry products. Due to the risk of resistance development and its transfer from animals to humans, the Belgian Royal Decree concerning the eradication of
Toine F H Bovee et al.
Mutation research, 778, 1-10 (2015-05-30)
It has previously been demonstrated by others that acetone extracts of Senecio jacobaea (syn. Jacobaea vulgaris, common or tansy ragwort) test positive in the Salmonella/microsome mutagenicity test (Ames test). Pyrrolizidine alkaloids (PAs) are thought to be responsible for these mutagenic
Mehrnoosh Shirangi et al.
Pharmaceutical research, 32(9), 3044-3054 (2015-04-03)
Polyesters with hydrophilic domains, i.e., poly(D,L-lactic-co-glycolic-co-hydroxymethyl glycolic acid) (PLGHMGA) and a multiblock copolymer of poly(ε-caprolactone)-PEG-poly(ε-caprolactone) and poly(L-lactide) ((PC-PEG-PC)-(PL)) are expected to cause less acylation of encapsulated peptides than fully hydrophobic matrices. Our purpose is to assess the extent and sites

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service