Skip to Content
Merck
All Photos(1)

Key Documents

V000697

Sigma-Aldrich

Magnesium perchlorate

anhydrous, p.a., 10-20 mesh

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(ClO4)2
CAS Number:
Molecular Weight:
223.21
EC Number:
MDL number:
PubChem Substance ID:

grade

anhydrous
p.a.

particle size

10-20 mesh

SMILES string

[Mg++].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O

InChI

1S/2ClHO4.Mg/c2*2-1(3,4)5;/h2*(H,2,3,4,5);/q;;+2/p-2

InChI key

MPCRDALPQLDDFX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey A Therrien et al.
Inorganic chemistry, 53(24), 12962-12972 (2014-10-23)
A series of pyridine- and lutidine-linked bis-N-heterocyclic carbene (NHC) palladium pincer complexes were electrochemically characterized and screened for CO2 reduction capability with 2,2,2-trifluoroethanol, acetic acid, or 2,2,2-trifluoroacetic acid (TFA) as proton sources. The lutidine-linked pincer complexes electrocatalytically reduce CO2 to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service