Skip to Content
Merck
All Photos(1)

Key Documents

1724780

USP

Zinc undecylenate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Zinc-10-undecenoate

Sign Into View Organizational & Contract Pricing

Select a Size

200 MG
CHF 365.00

CHF 365.00


Estimated to ship on26 April 2025


Request a Bulk Order

Select a Size

Change View
200 MG
CHF 365.00

About This Item

Empirical Formula (Hill Notation):
C22H38O4Zn
CAS Number:
Molecular Weight:
431.92
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

CHF 365.00


Estimated to ship on26 April 2025


Request a Bulk Order

API family

zinc undecylenate

packaging

pkg of 200 mg

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Zn](OC(=O)CCCCCCCCC=C)OC(=O)CCCCCCCCC=C

InChI

1S/2C11H20O2.Zn/c2*1-2-3-4-5-6-7-8-9-10-11(12)13;/h2*2H,1,3-10H2,(H,12,13);/q;;+2/p-2

InChI key

YMCOHQVWOBMDCZ-UHFFFAOYSA-L

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Zinc undecylenate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service