Skip to Content
Merck
All Photos(1)

Key Documents

1651621

USP

Delta-9-Tetrahydrocannabinol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Δ9-Tetrahydrocannabinol solution, Δ1-Tetrahydrocannabinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

cannabis

manufacturer/tradename

USP

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12CCC(C)=C[C@@]1([H])c3c(O)cc(CCCCC)cc3OC2(C)C

InChI

1S/C21H30O2/c1-5-6-7-8-15-12-18(22)20-16-11-14(2)9-10-17(16)21(3,4)23-19(20)13-15/h11-13,16-17,22H,5-10H2,1-4H3/t16-,17-/m1/s1

InChI key

CYQFCXCEBYINGO-IAGOWNOFSA-N

Gene Information

human ... CNR1(1268)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Delta-9-Tetrahydrocannabinol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Dronabinol Capsules
  • Dronabinol

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.70 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

1–2 of 2 Questions  
  1. What is the strength of this product?

    1 answer
    1. This Reference Standard is provided at a concentration of 1 mg/mL solution in methanol. USP Reference Standards are not intended for use as drugs, dietary supplements, or as medical devices. For further information and support please refer to the website of the issuing Pharmacopoeia.

      Helpful?

  2. Is this delta9-THC prepared from a phyto source or chemically synthesized?

    1 answer
    1. This is a product of the United States Pharmacopeia. The information available on the USP website states that this material is produced by chemical synthesis. Please see the link below to review the supplier web page in the Product Information Section located at the lower half of the listing:
      https://store.usp.org/product/1651621?_gl=1*zqqwy5*_gcl_au*NDA2MDgyNjYuMTY5Njk2MTEyMA..*_ga*MTg3MzY4OTg2Ny4xNjg2NTg2MTE5*_ga_DTGQ04CR27*MTcwMzcxMjI4My4xMS4xLjE3MDM3MTIyOTkuMC4wLjA.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service