Skip to Content
Merck
All Photos(1)

Key Documents

1409604

USP

Methenamine mandelate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Hexamethylenetetramine mandelate salt, 1,3,5,7-Tetraazatricyclo[3.3.1.13,7]decane, Hexamine, Methenamine

Sign Into View Organizational & Contract Pricing

Select a Size

200 MG
CHF 395.00

CHF 395.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
200 MG
CHF 395.00

About This Item

Empirical Formula (Hill Notation):
C6H12N4 · C8H8O3
CAS Number:
Molecular Weight:
292.33
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

CHF 395.00


Estimated to ship on22 May 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

methenamine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

C1N2CN3CN1CN(C2)C3.OC(C(O)=O)c4ccccc4

InChI

1S/C8H8O3.C6H12N4/c9-7(8(10)11)6-4-2-1-3-5-6;1-7-2-9-4-8(1)5-10(3-7)6-9/h1-5,7,9H,(H,10,11);1-6H2

InChI key

UXNFIJPHRQEWRQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Methenamine mandelate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Methenamine Mandelate Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

482.0 °F - closed cup

Flash Point(C)

250 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service