Skip to Content
Merck
All Photos(1)

Key Documents

1238578

USP

6-D-Cys EptifIbatide

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N6-Amidino-N2-(3-mercaptopropionyl)-L-lysylglycyl-L-alpha-aspartyl-L-tryptophyl-L-prolyl-D-cysteinamide, cyclic(1-6)-disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H49N11O9S2
Molecular Weight:
831.96
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

API family

eptifibatide

packaging

pkg of 10 mg

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

This product is part of the USP Biologics program.
Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service