Skip to Content
Merck
All Photos(1)

Key Documents

SY3H3D676DFE

Synthonix Corporation

Bromotris(dimethylamino)phosphonium Hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
388.07
MDL number:

Iupac Name

bromo[tris(dimethylamino)]phosphonium hexafluorophosphate

Inchi Code

1S/C6H18BrN3P.F6P/c1-8(2)11(7,9(3)4)10(5)6;1-7(2,3,4,5)6/h1-6H3;/q+1;-1

InChI key

XELPBWPBGHCIKX-UHFFFAOYSA-N

storage temp.

Room Temperature

Purity

98%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Solid

Product Link

https://synthonix.com/41626.html

Pictograms

CorrosionHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://synthonix.com/41626.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library