Skip to Content
Merck
All Photos(1)

Key Documents

SY3371476532

Synthonix Corporation

3-Cyanoazetidine-1-sulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
180.61
MDL number:

Iupac Name

3-cyanoazetidine-1-sulfonyl chloride

Inchi Code

1S/C4H5ClN2O2S/c5-10(8,9)7-2-4(1-6)3-7/h4H,2-3H2

InChI key

MHBMAFUVWZCQBB-UHFFFAOYSA-N

storage temp.

Refrigerator

Purity

0.97

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Solid

Product Link

https://synthonix.com/7013.html

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

MSDS

https://synthonix.com/7013.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library