Skip to Content
Merck
All Photos(1)

Documents

Z227021

Spherisorb® NH2 Guard Cartridge

5 μm particle size, L × I.D. 1 cm × 4.6 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Agency

suitable for USP L8

feature

endcapped: no

packaging

pkg of 4 ea

extent of labeling

1.9% Carbon loading

technique(s)

HPLC: suitable

L × I.D.

1 cm × 4.6 mm

matrix active group

amino phase

particle size

5 μm

compatibility

for use with 3.0 to 4.6 mm column

separation technique

anion exchange
hydrophilic interaction (HILIC)
normal phase

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Spherisorb is a registered trademark of Waters

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service