Skip to Content
Merck
All Photos(1)

Key Documents

SAAR5871061

Stannous chloride

anhydrous, univAR

Synonym(s):

Tin(II) chloride, Stannous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SnCl2
CAS Number:
Molecular Weight:
189.62
MDL number:
UNSPSC Code:
12352302

grade

anhydrous

product line

univAR

packaging

poly bottle of 12 × 500 g (SAAR5871061EM)

bp

652 °C (lit.)

mp

246 °C (lit.)

storage temp.

2-30°C

SMILES string

Cl[SnH2]Cl

InChI

1S/2ClH.Sn/h2*1H;/q;;+2/p-2

InChI key

AXZWODMDQAVCJE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Analysed reagent
For laboratory use only

Legal Information

univAR is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Cardio-vascular system, Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service