Skip to Content
Merck
All Photos(1)

Key Documents

SAAR5046820

Potassium permanganate

univAR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KMnO4
CAS Number:
Molecular Weight:
158.03
MDL number:
UNSPSC Code:
12352312
grade:
reagent grade
Pricing and availability is not currently available.

grade

reagent grade

product line

univAR

packaging

poly bottle of 1 × 500 g (SAAR5046820EM)

storage temp.

2-30°C

SMILES string

[K+].[O-][Mn](=O)(=O)=O

InChI

1S/K.Mn.4O/q+1;;;;;-1

InChI key

VZJVWSHVAAUDKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Analysed reagent
For laboratory use only
Suitable for heterogeneous oxidations.

Legal Information

univAR is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 2 - Repr. 2 - Skin Corr. 1 - STOT RE 2 Inhalation

Target Organs

Brain

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service