Skip to Content
Merck
All Photos(1)

Key Documents

PS403

Metolachlor

analytical standard

Synonym(s):

2-Chloro-N-(2-ethyl-6-methylphenyl)-N-(-2-methoxy-1-methylethyl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22ClNO2
CAS Number:
Molecular Weight:
283.79
Beilstein:
2743537
EC Number:
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. PS-403

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCc1cccc(C)c1N(C(C)COC)C(=O)CCl

InChI

1S/C15H22ClNO2/c1-5-13-8-6-7-11(2)15(13)17(14(18)9-16)12(3)10-19-4/h6-8,12H,5,9-10H2,1-4H3

InChI key

WVQBLGZPHOPPFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zisis Vryzas et al.
Chemosphere, 89(11), 1330-1338 (2012-06-29)
Biotransformation studies of atrazine, metolachlor and evolution of their metabolites were carried out in soils and subsoils of Northern Greece. Trace atrazine, its metabolites and metolachlor residues were detected in field soil samples 1 year after their application. The biotransformation
Sean Hartnett et al.
Chemosphere, 90(3), 1258-1266 (2012-10-23)
Metolachlor is one of the most commonly used herbicides in the United States. Protein synthesis is inhibited when roots and shoots of susceptible plants absorb this synthetic herbicide. While quite effective in killing weeds, several studies have shown that exposure
J Restivo et al.
Journal of hazardous materials, 239-240, 249-256 (2012-09-27)
The catalytic ozonation of the herbicide metolachlor (MTLC) was tested using carbon nanomaterials as catalysts. Multiwalled carbon nanotubes were used in semi-batch experiments and carbon nanofibres grown on a honeycomb cordierite monolith were tested in continuous experiments. The application of
Richard E Lizotte et al.
The Science of the total environment, 427-428, 373-381 (2012-05-09)
We examined the mitigation efficiency of a managed riverine wetland amended with a mixture of suspended sediment, two nutrients (nitrogen and phosphorus), and three pesticides (atrazine, metolachlor, and permethrin) during a simulated agricultural runoff event. Hydrologic management of the 500
Jianfeng Wang et al.
Journal of chromatography. A, 1247, 10-17 (2012-06-12)
A liquid-phase microextraction (LPME) methodology based on the use of porous polyvinylidene fluoride (PVDF) hollow fibres was developed for extracting seven pesticides from cucumbers. The seven pesticides include propoxur, carbofuran, atrazine, cyanatryn, metolachlor, prometryn and tebuconazole. The PVDF hollow fibre

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service