Skip to Content
Merck
All Photos(1)

Key Documents

PHR9066

Supelco

10-Bromo-5H-dibenz[b,f]azepine-5-carboxamide

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Carbamazepine impurity G (PhEur), 10-Bromocarbamazepine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 720.00

CHF 720.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
CHF 720.00

About This Item

Empirical Formula (Hill Notation):
C15H11BrN2O
CAS Number:
Molecular Weight:
315.16
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

CHF 720.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

application(s)

pharmaceutical small molecule

storage temp.

2-8°C

SMILES string

BrC1=Cc2c(cccc2)N(c3c1cccc3)C(=O)N

InChI

1S/C15H11BrN2O/c16-12-9-10-5-1-3-7-13(10)18(15(17)19)14-8-4-2-6-11(12)14/h1-9H,(H2,17,19)

InChI key

FGWUKVYDIGVLTD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034.
This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma aldrich.com

Application

These Reference Materials are suitable for use in several analytical applications including, but not limited to, method development for qualitative and quantitative analyses, daily calibration, and routine quality control testing.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1A - Resp. Sens. 1A - Skin Sens. 1A - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service