Skip to Content
Merck
All Photos(1)

Documents

PHL80430

Dihydrokaempferol

phyproof® Reference Substance

Synonym(s):

(+)-Aromadendrol, (+)-Dihydrokaempferol, (2R,3R)-3,4′,5,7-Tetrahydroxyflavanone, (2R,3R)-3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-2,3-dihydro-4H-chromen-4-one, Aromadendrin, Katuranin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O6
CAS Number:
Molecular Weight:
288.25
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

SMILES string

O[C@@H]1[C@H](Oc2cc(O)cc(O)c2C1=O)c3ccc(O)cc3

InChI

1S/C15H12O6/c16-8-3-1-7(2-4-8)15-14(20)13(19)12-10(18)5-9(17)6-11(12)21-15/h1-6,14-18,20H/t14-,15+/m0/s1

InChI key

PADQINQHPQKXNL-LSDHHAIUSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Other Notes

This compound is commonly found in plants of the genus: ginkgo silybum thymus

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service