Skip to Content
Merck
All Photos(1)

Documents

G006116

UHPLC/MS Analysis of Heroin and Related Compounds on Titan C18

application for UHPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

UHPLC: suitable

test parameters

column: Titan C18 5 cm x 2.1 mm I.D., 1.9 μm particles (577122-U)
mobile phase: [A] 5 mM ammonium acetate in 95:5, water:acetonitrile; [B] 5 mM ammonium acetate in 5:95, water:acetonitrile
gradient: 0 to 100% B in 3 min, held at 100% B for 1 min
flow rate: 0.6 mL/min
pressure: 6770 psi (467 bar)
column temp.: 35 °C
detector: MS-TOF, XIC
injection: 0.5 μL
sample: 1 μg/mL in 95:5, water:methanol

suitability

application for UHPLC

application(s)

forensics and toxicology

Analysis Note

This method shows heroin and related compounds separated on a Titan C18 column and MS detection. The highest quality LC-MS solvents were used to supply low background interference and low particulate contaminants for robust, trouble-free operation. Cerilliant CRMs provided reliable identification and quantification.

Legal Information

Analyte

Description

    Morphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    Procaine hydrochloride, VETRANAL®, analytical standard
    Codeine solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    Heroin solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
    Alprazolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    Papaverine hydrochloride, powder
    Flunitrazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    Diazepam solution, 1.0 mg/mL in methanol, analytical standard, for drug analysis
    Noscapine hydrochloride hydrate, analytical standard
    See All (9)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service