Skip to Content
Merck
All Photos(1)

Key Documents

G005724

HPLC Analysis of Proteins on BIOshell A160 Peptide C18

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: BIOshell A160 Peptide C18, 15 cm x 2.1 mm I.D., 2.7 μm particles (66905-U)
mobile phase: [A] 75:25, 0.1% TFA in water: 0.08% TFA in acetonitrile; [B] 25:75, 0.1% TFA in water: 0.08% TFA in acetonitrile
gradient: held at 0% B for 0.5 min; 0 to 100% B in 7 min
flow rate: 0.3 mL/min
pressure: 4743 psi (327 bar)
column temp.: 35 °C
detector: UV, 220 nm
injection: 1 μL
sample: 0.2 - 0.6 g/L in mobile phase A

suitability

application for HPLC

application(s)

life science and biopharma

Legal Information

Analyte

Description

    Ribonuclease A from bovine pancreas, Type I-A, powder, ≥60% RNase A basis (SDS-PAGE), ≥50 Kunitz units/mg protein
    Adrenomedullin rat, ≥95% (HPLC)
    Cytochrome c from chicken heart, ≥95% based on Mol. Wt. 12,222 basis
    Lysozyme from chicken egg white, powder or granules, ≥39,000 units/mg protein
    Superoxide Dismutase from bovine erythrocytes, BioReagent, ≥3,000 units/mg protein, suitable for cell culture, lyophilized powder
    Trypsin Inhibitor from Glycine max (soybean), BioUltra, lyophilized powder, ≥95% (Kunitz inhibitor, SDS-PAGE)
    Carbonic Anhydrase from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
    Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
    See All (8)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service