97148
Cyflufenamid E-isomer
PESTANAL®, analytical standard
Synonym(s):
(E)-N-[α-(Cyclopropylmethoxyimino)-2,3-difluoro-6-(trifluoromethyl)benzyl]-2-phenylacetamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C20H17F5N2O2
Molecular Weight:
412.35
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
analytical standard
Quality Level
product line
PESTANAL®
Assay
≥98.0% (HPLC)
format
neat
storage temp.
−20°C
Related Categories
General description
Cyflufenamid is a preventative and curative amidoxine fungicide for the control of powdery mildew in cereals and other crops. It has a low aqueous solubility and is non-volatile. It is approved according Reg. (EC) No 1107/2009 for its use as a plant protection product in the European Union (EU). Maximim residue levels (MRLs) have been set for cyflufenamid (sum of cyflufenamid Z- and its E-isomer), expressed as cyflufenamid according to Reg. (EU) 2022/476 in various products of plant and animal origin in the range of 0.01 - 0.2 mg/kg
Application
Environmental
Legal Information
PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service