Skip to Content
Merck
All Photos(1)

Key Documents

96606

Supelco

1-Vinylimidazole

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2
CAS Number:
Molecular Weight:
94.11
Beilstein:
105321
MDL number:
UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable

refractive index

n20/D 1.533 (lit.)

bp

192-194 °C (lit.)
78-79 °C/13 mmHg (lit.)

density

1.039 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

C=Cn1ccnc1

InChI

1S/C5H6N2/c1-2-7-4-3-6-5-7/h2-5H,1H2

InChI key

OSSNTDFYBPYIEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

183.2 °F - Pensky-Martens closed cup

Flash Point(C)

84 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service