Skip to Content
Merck
All Photos(1)

Key Documents

845122

TOYOPEARL® Phenyl-650S MiniChrom Column

phase phenyl, 35 μm particle size, pore size 1000 Å, L × I.D. 10 cm × 8 mm, MiniChrom Column

Synonym(s):

MiniChrom

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500

material

PEEK

product line

TOYOPEARL®

analyte chemical class(es)

proteins

manufacturer/tradename

Tosoh 45122

parameter

≤50% organic solvent
3 bar max. pressure (resin)
30 bar max. pressure (column)

technique(s)

HPLC: suitable
LPLC: suitable

L × I.D.

10 cm × 8 mm

bed volume

5 mL

matrix

hydroxylated methacrylic polymer

matrix active group

phenyl phase

particle size

35 μm

pore size

1000 Å

operating pH

1-13

capacity

40 g/L±10 adsorption capacity (lysozyme)

separation technique

hydrophobic interaction (HIC)

Looking for similar products? Visit Product Comparison Guide

General description

MiniChrom columns for TOYOPEARL® and TSKgel® are pre-packed with the most popular ion exchange, mixed-mode, hydrophobic interaction or affinity media. These columns are available in 5 mL volume, 8 mm ID x 10 cm L, and can be operated with standard laboratory LC systems. MiniChrom columns for TOYOPEARL and TSKgel provide:
  • Direct connection to common laboratory LC Systems
  • Easy parameter screening and method optimization
  • Fast small scale purification
  • Reliable and reproducible results

Compatibility

Columns are tolerant to aqueous buffers and salt solutions, 1 mol/L alkaline solutions, 1 mol/L non-oxidizing mineral acids, 8 mol/L urea, 6 mol/L guanidine hydrochloride, non-halogenated organic solvents and detergents. They are not compatible with strong oxidants and halogenated solvents.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

Shipped in 20% (v/v) ethanol, 0.15 M NaCl.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation
Toyopearl is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service