Skip to Content
Merck
All Photos(1)

Key Documents

819805

TOYOPEARL® Ether-650M Bulk Media

bottle of 25 mL, phase ether, 65 μm particle size

Synonym(s):

TOYOPEARL® Ether-650 HIC Bulk Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
Pricing and availability is not currently available.

product line

TOYOPEARL®

form

slurry

packaging

bottle of 25 mL

manufacturer/tradename

Tosoh 19805

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

ether phase

particle size

65 μm

operating pH

1-13

capacity

20 g/L±10 adsorption capacity (lysozyme)

separation technique

hydrophobic interaction (HIC)

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl Ether-650 resin has the highest hydrophilicity of the HIC ligands offered by Tosoh Bioscience. It is useful for very hydrophobic molecules such as membrane proteins. It is typically used in intermediate purification and polishing steps. For high performance separations, the use of TSKgel Ether-5PW resin is suggested. Both products have the same selectivity.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service