Skip to Content
Merck
All Photos(1)

Documents

78340

Supelco

1-Octene

reference material

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH=CH2
CAS Number:
Molecular Weight:
112.21
Beilstein:
1734497
MDL number:
UNSPSC Code:
41116107

grade

reference material

vapor density

3.9 (vs air)

vapor pressure

36 mmHg ( 38 °C)

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

446 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

3.9 %

refractive index

n20/D 1.408 (lit.)
n20/D 1.409 (lit.)

bp

122-123 °C (lit.)

mp

−101 °C (lit.)

density

0.715 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

CCCCCCC=C

InChI

1S/C8H16/c1-3-5-7-8-6-4-2/h3H,1,4-8H2,2H3

InChI key

KWKAKUADMBZCLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service