Key Documents
56728
Supelco
Stainless Steel 1/16 in. Capillary Tubing
L × O.D. × I.D. 30 cm × 1/16 in. × 0.020 in.
About This Item
UNSPSC Code:
41115700
eCl@ss:
32119290
Pricing and availability is not currently available.
L × O.D. × I.D.
30 cm × 1/16 in. × 0.020 in.
color
yellow band
General description
316 stainless steel, 1/16 in. O.D. 1 m length is coiled, all other dimensions are straight.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)