Skip to Content
Merck
All Photos(1)

Documents

54919

Supelco

Nucleosil® C18 HPLC Column

5 μm particle size, L × I.D. 25 cm × 3.2 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

Agency

suitable for USP L1

feature

endcapped

manufacturer/tradename

Nucleosil®

extent of labeling

15% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

25 cm × 3.2 mm

surface area

350 m2/g

matrix

octadecyl

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

100 Å

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Nucleosil® C18 HPLC Column features nonpolar, endcapped phases for reversed-phase chromatography. The separation mechanism is based on hydrophobic (van der Waals) interactions and slight residual silanol interactions.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Nucleosil is a registered trademark of Macherey-Nagel GmbH & Co. KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service