Skip to Content
Merck
All Photos(1)

Documents

54479

Rheodyne® Model 7000L Stream Switching Valve, Large Bore

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854

material

stainless steel valve

feature

manual
number of ports 6
positions 2

manufacturer/tradename

Rheodyne 7000L

parameter

5000 psi max. pressure (350 bar) (factory set)

fitting

10-32 Rheodyne thread

General description

The 2-position 7000L Stream Switching Valve has a flow passage of 1 mm (0.040″).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Rheodyne is a registered trademark of IDEX Health & Science, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service