Skip to Content
Merck
All Photos(1)

Documents

48549

Supelco

4-Nitrophenol

Synonym(s):

p-Nitrophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4OH
CAS Number:
Molecular Weight:
139.11
Beilstein:
1281877
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor pressure

0.6 mmHg ( 120 °C)

CofA

current certificate can be downloaded

autoignition temp.

541 °F

packaging

ampule of 5000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

279 °C (lit.)

mp

110-115 °C (lit.)

solubility

H2O: soluble 15 g/L

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

O=N(C1=CC=C(O)C=C1)=O

InChI

1S/C6H5NO3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H

InChI key

BTJIUGUIPKRLHP-UHFFFAOYSA-N

Gene Information

human ... UGT1A4(54657)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - STOT RE 2 Oral

Target Organs

Kidney,Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

336.2 °F - closed cup

Flash Point(C)

169.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali Mehdinia et al.
Journal of chromatography. A, 1283, 82-88 (2013-03-08)
In this study, an analytical procedure for the selective extraction and detection of 4-nitrophenol (4-NP) was investigated by using of molecularly imprinted polymer on the surface of magnetic nanoparticles (MNPs). The magnetic nanoparticles were modified by tetraethyl orthosilicate (TEOS) and
Ipsita K Sen et al.
Carbohydrate polymers, 91(2), 518-528 (2012-11-06)
Gold nanoparticles were synthesized by reducing chloroauric acid with a glucan, isolated from an edible mushroom Pleurotus florida, cultivar Assam Florida. Here, glucan acts as reducing as well as stabilizing agent. The synthesized gold nanoparticles were characterized by UV-visible spectroscopy
Alex V Birk et al.
The Journal of laboratory and clinical medicine, 140(3), 166-175 (2002-09-25)
Extracellular adenosine triphosphate (ATP) regulates platelet reactivity by way of direct action on platelet purinergic receptors or by hydrolysis to adenosine diphosphate (ADP). Subsequent metabolism of ATP and ADP to adenosine monophosphate (AMP) and adenosine inhibits platelet aggregation. Endothelial cell
Simon De Corte et al.
Colloids and surfaces. B, Biointerfaces, 102, 898-904 (2012-10-31)
An increasing demand for catalytic Pd nanoparticles has motivated the search for sustainable production methods. An innovative approach uses bacterial cells as support material for synthesizing Pd nanoparticles by reduction of Pd(II) with e.g. hydrogen or formate. Nevertheless, drawbacks of
Jau-Rung Chiou et al.
Journal of hazardous materials, 248-249, 394-400 (2013-02-19)
Silver/iron oxide composite nanoparticles have been synthesized successfully via a facile one-pot green route by the use of l-arginine, which created an aqueous solution of about pH 10 and acted as a reducing agent for the successive formation of iron

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service