Skip to Content
Merck
All Photos(1)

Documents

48079

Supelco

Perylene solution

certified reference material, 2000 μg/mL in dichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL in dichloromethane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C20H12/c1-5-13-6-2-11-17-18-12-4-8-14-7-3-10-16(20(14)18)15(9-1)19(13)17/h1-12H

InChI key

CSHWQDPOILHKBI-UHFFFAOYSA-N

General description

Perylene is a carcinogenic compound, belonging to the class of polycyclic aromatic hydrocarbons, widely distributed in the environment from different sources.

Application

Perylene solution may be used as a certified reference standard for the quantification of the analyte in soil samples using gas chromatography coupled to mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assessment of polycyclic aromatic hydrocarbons (PAHs) in soil of a Natural Reserve (Isola delle Femmine)(Italy) located in front of a plant for the production of cement
Orecchio S
Journal of Hazardous Materials, 173(1-3), 358-368 (2010)
Determination of polynuclear aromatic hydrocarbons in water by flotation enrichment and HPLC
Bo-Xing X and Yu-Zhi F
Talanta, 35(11), 891-894 (1988)
Maurus Biedermann et al.
Journal of chromatography. A, 1281, 106-114 (2013-02-12)
Insertion of a programmed temperature vaporizing (PTV) injector under conditions of concurrent solvent recondensation (CSR) into the on-line HPLC-GC interface for on-column transfer (such as the retention gap technique with partially concurrent eluent evaporation) enables filtering off high boiling or
Fei Ma et al.
Bioorganic & medicinal chemistry letters, 23(6), 1689-1692 (2013-02-16)
The complex of Hypocrellin A with Al(3+) is prepared in water solution by a facile method. The water-solubility and stability of complexes are improved. Irradiation of Al(3+)-HA complex results in higher efficient generation of singlet oxygen ((1)O2) and photocleavage ability
Angel Trigos et al.
Natural product communications, 8(2), 187-189 (2013-03-22)
From cultures of Cercospora piaropi, a phytopathogenic fungus isolated from symptomatic leaves of water hyacinth was obtained a red compound, which, according to the spectroscopic data, was epi-cercosporin. It showed in vitro antiproliferative activity against the panel of human solid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service