Skip to Content
Merck
All Photos(1)

Key Documents

46933

Supelco

Citric acid

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOH)(CH2COOH)2
CAS Number:
Molecular Weight:
192.12
Beilstein:
782061
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
E Number:
E330

grade

analytical standard

Assay

99.5% (as is basis)

CofA

current certificate can be downloaded

expl. lim.

8 %, 65 °F

packaging

pkg of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

pKa 

(1) 3.13, (2) 4.76, (3) 6.4

mp

153-159 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O

InChI

1S/C6H8O7/c7-3(8)1-6(13,5(11)12)2-4(9)10/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

KRKNYBCHXYNGOX-UHFFFAOYSA-N

Gene Information

human ... SRC(6714)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria José Garcia-Fernandez et al.
Biofouling, 29(3), 261-271 (2013-02-27)
Prevention and management of wound infections receive a lot of attention, since the presence of micro-organisms interferes with the wound-healing process. The aim of this work was to use cyclodextrins (CDs) to endow hydrogels and gauzes with the ability to
Wayne B Batchelor et al.
Circulation, 106(12), 1470-1476 (2002-09-18)
The relative anti-aggregatory effects of currently prescribed platelet glycoprotein IIb/IIIa receptor antagonists during and after percutaneous coronary intervention for acute coronary syndromes have not been established. We randomized 70 acute coronary syndrome patients undergoing percutaneous coronary intervention to receive abciximab
Benjamin Nota et al.
American journal of human genetics, 92(4), 627-631 (2013-04-09)
The Krebs cycle is of fundamental importance for the generation of the energetic and molecular needs of both prokaryotic and eukaryotic cells. Both enantiomers of metabolite 2-hydroxyglutarate are directly linked to this pivotal biochemical pathway and are found elevated not
Konrad Hoetzenecker et al.
European heart journal, 36(11), 676-685 (2013-01-17)
Supernatants of serum-free cultured mononuclear cells (MNC) contain a mix of immunomodulating factors (secretome), which have been shown to attenuate detrimental inflammatory responses following myocardial ischaemia. Inflammatory dilated cardiomyopathy (iDCM) is a common cause of heart failure in young patients.
Tetsuji Sawada et al.
Gastric cancer : official journal of the International Gastric Cancer Association and the Japanese Gastric Cancer Association, 18(1), 119-128 (2014-02-04)
The G-Project committee was erected by the Japan Society for Gastroenterological Carcinogenesis with an aim of establishing a new classification scheme based on molecular biological characteristics that would supplement the conventional TNM classification to better predict outcome. In a literature

Protocols

Separation of Fumaric acid; Oxalic acid; Citric acid

Science Slam panel: Leading gene therapy developers discuss commercialization challenges and the importance of robust process development plans.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service