Skip to Content
Merck
All Photos(1)

Key Documents

442880

Supelco

2,4-Dichlorophenol-d3

analytical standard

Synonym(s):

2,4-Dichlorophenol-3,5,6-d3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6D3OH
CAS Number:
Molecular Weight:
166.02
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 50 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

209-210 °C (lit.)

mp

42-43 °C (lit.)

application(s)

agriculture
environmental

format

neat

mass shift

M+3

storage temp.

room temp

SMILES string

[2H]c1c([2H])c(Cl)c([2H])c(Cl)c1O

InChI

1S/C6H4Cl2O/c7-4-1-2-6(9)5(8)3-4/h1-3,9H/i1D,2D,3D

InChI key

HFZWRUODUSTPEG-CBYSEHNBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

237.2 °F - closed cup

Flash Point(C)

114 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service