Skip to Content
Merck
All Photos(1)

Key Documents

442434

Supelco

Acetaldehyde-2,4-DNPH

analytical standard

Synonym(s):

Acetaldehyde-2,4-dinitrophenylhydrazone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
224.17
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

165-168 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

C\C=N\Nc1ccc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C8H8N4O4/c1-2-9-10-7-4-3-6(11(13)14)5-8(7)12(15)16/h2-5,10H,1H3/b9-2+

InChI key

ONBOQRNOMHHDFB-XNWCZRBMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetaldehyde-2,4-DNPH can be obtained as an oxidation product of the reaction between alpha-pinene with hydroxyl radicals, which is monitored by gas chromatography, high performance liquid chromatography and detected by using mass spectral analysis.

Application

Acetaldehyde-2,4-DNPH has been used as an internal standard to study the ozonation of an antidepressant fluoxetine (FX) and its principle metabolite product norfluoxetine (NFX) using kinetic method of analysis and high performance liquid chromatography coupled with mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ozonation of antidepressant fluoxetine and its metabolite product norfluoxetine: Kinetics, intermediates and toxicity
Zhao Y, et al.
Chemical Engineering Journal, 316, 951-963 (2017)
Identification of the oxidation products of the reaction between ?-pinene and hydroxyl radicals by gas and high-performance liquid chromatography with mass spectrometric detection
Bergh DVV, et al.
Journal of Chromatography A, 896, 135-148 (2000)
Denise Biagini et al.
Talanta, 216, 120979-120979 (2020-05-28)
Biomarkers of oxidative stress are generally measured in blood and its derivatives. However, the invasiveness of blood collection makes the monitoring of such chemicals during exercise not feasible. Saliva analysis is an interesting approach in sport medicine because the collection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service