28865-U
Super Clean (In-Line Design) Gas Purifier
triple trap (hydrocarbon, moisture, oxygen), gas specific for helium, stainless steel
About This Item
Recommended Products
material
stainless steel
Looking for similar products? Visit Product Comparison Guide
General description
Note: Does not include Click-on Connectors, these must be purchased before initial use.
Dimensions:
- Cartridge and Connectors: 11 1/2 in. long x 1 1/4 in. diameter
- Cartridge Only: 7 5/8 in. long x 1 1/4 in. diameter
accessory
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service