Skip to Content
Merck
All Photos(3)

Key Documents

28517-U

Supelco

ASSET® EZ4-ICA Dry Sampler

for sampling isocyanic acid (ICA), for use with for use with LC-MS or LC-MS/MS, pkg of 10 ea, ASSET®

Synonym(s):

ASSET® EZ4 Dry Samplers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12190000
NACRES:
NB.24
Pricing and availability is not currently available.

Product Name

ASSET® EZ4-ICA Dry Sampler, pkg of 10 ea

product line

ASSET®

packaging

pkg of 10 ea

technique(s)

active air sampling: suitable

matrix

DBA-Impregnated Glass Fiber Filter

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for sampling isocyanic acid (ICA)
for use with for use with LC-MS or LC-MS/MS

Looking for similar products? Visit Product Comparison Guide

Storage and Stability

Store at 4 °C. (Refrigerate before use)

Legal Information

Asset is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B Inhalation - Eye Dam. 1 - Skin Corr. 1A - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Gylestam et al.
The Annals of occupational hygiene, 58(3), 340-354 (2013-12-29)
An advanced design of a denuder impactor (DI) sampler has been developed for characterization of possible airborne isocyanate exposure in different particle size fractions. The sampler is equipped with 12 different parallel denuder tubes, 4 impaction stages with the cut-off

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service