Key Documents
27401
Supelco
Screw cap
black polypropylene hole cap,
large opening in top of cap, no septum
, pkg of 100 ea, thread for 8-425
About This Item
UNSPSC Code:
41122001
material
black polypropylene hole cap
packaging
pkg of 100 ea
fitting
thread for 8-425
General description
Screw caps (large, opening in top of cap, no septum, thread 8-425) are available as hole caps for use with sealing septa. Polypropylene caps can withstand a maximum temperature of 135°C.
Choose from one of the most recent versions:
Certificates of Analysis (COA)