Key Documents
23393-U
Supelco
Stainless Steel Tension Springs for TD Tubes
greener alternative
About This Item
UNSPSC Code:
41104008
NACRES:
NB.24
Pricing and availability is not currently available.
packaging
pack of 100 ea
greener alternative product characteristics
Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
technique(s)
active air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
greener alternative category
General description
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. Thermal desorption tubes are reusable at least 50 to 100 times, and reduces the usage of accessories like transfer vials, caps, and pipette tips, thus aligns with “Waste Prevention”. Also, these tubes eliminate the usage of toxic solvents like CS2 to extract the VOCs of interest, and thus aligns with “Safer Solvents and Auxiliaries” principle. Click here for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)