Documents
22492
Supelco
Supeltex M-4 Packed Column Ferrule, 1/4 in. Column O.D.
I.D. 1/4 in., 1/4 in. O.D. Column, pkg of 10 ea
About This Item
UNSPSC Code:
41121815
packaging
pkg of 10 ea
I.D.
1/4 in.
compatibility
1/4 in. O.D. Column
General description
Ferrules are suitable for the attachment of the column and other glass components to the system. Flexible graphite ferrules withstand high temperatures and are capable of compression.
Supelco has refined the design of these graphite ferrules to provide a clean, sharp profile with minimal flash.
Characteristics:
Seals at one-quarter turn past fingertight. Maximum sealing surface contact, reduced risk of column contamination at installation.
Material Composition: flexible graphite
Maximum Temperature: 450 °C
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Walter Jennings
Gas Chromatography with Glass Capillary Columns (2012)