Key Documents
22423
Supelco
In-Line Pressure Gauge
pressure range 0-100 psi
Synonym(s):
PRESSURE GAUGE KIT FOR PURIFIER EA
About This Item
UNSPSC Code:
41112500
parameter
0-100 psi pressure range
General description
Install a pressure gauge in the gas delivery system to measure pressure at any given point. For example, install one upstream and downstream of the High Capacity Gas Purifier. When the pressure drop across the purifier exceeds 10 psi, the converter tube should be replaced. Contains:
  • 2 in. (5 cm) diameter steel and copper alloy gauge
  • NPT to Swagelok adapter
  • 1/8 in. tee
Nuts and ferrules included.
Choose from one of the most recent versions:
Certificates of Analysis (COA)