Key Documents
22327
Supelco
Stainless Steel Screening
pkg of 1 ea
About This Item
UNSPSC Code:
12000000
Pricing and availability is not currently available.
General description
An alternative to the use of wool plugs to hold packing material inside columns. Includes a single 2 in. x 2 in. (5 cm x 5 cm) sheet of 10 μm pore size material.
Choose from one of the most recent versions:
Certificates of Analysis (COA)