Skip to Content
Merck
All Photos(1)

Key Documents

13340-U

Supelco

Dowex Monosphere 650C UPW hydrogen form

bucket of 1000 g

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41115710
NACRES:
SB.52

description

cross-linkage 8%

form

beads

quality

purified 650C

packaging

bucket of 1000 g

parameter

150 °C max. temp.

moisture

46-51%

technique(s)

LPLC: suitable

matrix

styrene divinylbenzene (gel)

matrix active group

sulfonic acid functional group

particle size

21-25 mesh
600-700 μm std dev

operating pH

0-14

capacity

2.0 meq/mL by wetted bed volume

separation technique

cation exchange

InChI

1S/C10H12.C10H10.C8H8/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2

InChI key

NWUYHJFMYQTDRP-UHFFFAOYSA-N

General description

Dowex Monosphere 650C is a strong basic cation exchange gel resin with sulfonic acid as functional group. Esterification of acetic acid with propyl alcohol over heterogeneous catalyst, Dowex Monosphere 650C, has been reported.

Application

Dowex Monosphere 650C UPW hydrogen form packed column was used in flow based ammonia gas analyzer.
Strongly acidic cation exchange resin capable of demineralization for ultrapure water; meets requirements of FDA Food Additive Regulation 21 CFR 173.25.

Physical properties

Color: light to yellow brown
Odor: none to slight
Appearance: translucent to opaque fine solid bead

Legal Information

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetic Study of Esterification of Acetic Acid with n-butanol and isobutanol Catalyzed by Ion Exchange Resin
Toor, Amrit Pal, et al.
Bulletin of Chemical Reaction Engineering and Catalysis, 6, 23-30 (2011)
Shin-Ichi Ohira et al.
Talanta, 116, 527-534 (2013-10-24)
A robust and fully automated indoor ammonia gas monitoring system with an open channel scrubber (OCS) was developed. The sample gas channel dimensions, hydrophilic surface treatment to produce a thin absorbing solution layer, and solution flow rate of the OCS
Optimization of the reverse osmosis seawater demineralization technologies for a power producing industry
Fendri, Fredg, Tatyana Mitchenko, and Zakhar Maletskyi
Desalination and Water Treatment, 25, 84-90 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service