Skip to Content
Merck
All Photos(1)

Key Documents

00282

Supelco

Diheptyl phthalate

analytical standard

Synonym(s):

Phthalic acid diheptyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-[CO2(CH2)6CH3]2
CAS Number:
Molecular Weight:
362.50
Beilstein:
2336623
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.485 (lit.)

bp

195 °C/0.04 mmHg (lit.)

density

0.988 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCC

InChI

1S/C22H34O4/c1-3-5-7-9-13-17-25-21(23)19-15-11-12-16-20(19)22(24)26-18-14-10-8-6-4-2/h11-12,15-16H,3-10,13-14,17-18H2,1-2H3

InChI key

JQCXWCOOWVGKMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diheptyl phthalate belongs to the class of phthalate esters (PAEs), a group of commercially important chemical compounds used as an industrial solvent and plasticizer in polyvinyl chloride (PVC) plastics. PAEs are regarded as ubiquitous environmental contaminants which are toxic to living organisms.[1]

Application

Diheptyl phthalate may be used as an analytical reference standard for the determination of the analyte in water samples by magnetic solid-phase extraction based on magnetic multi-walled carbon nanotubes followed by gas chromatography-tandem mass spectrometry (GC-MS/MS)[2] and in food samples by liquid chromatography-tandem mass spectrometry (LC-MS/MS).[3]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of 23 phthalic acid esters in food by liquid chromatography tandem mass spectrometry
Xu D, et al.
Journal of Chromatography A, 1324(9), 49-56 (2014)
Determination of trace leaching phthalate esters in water by magnetic solid phase extraction based on magnetic multi-walled carbon nanotubes followed by GC-MS/MS
Jiao Y, et al.
Analytical Methods : Advancing Methods and Applications, 4(9), 2729-2734 (2012)
Fumiyuki Nakane et al.
The Journal of toxicological sciences, 37(3), 527-537 (2012-06-13)
The 26-week oral toxicity of diheptyl phthalate (DHP), a peroxisome proliferator-activated receptor α (PPARα) agonist, with special emphasis on the potential induction of hepatocellular proliferative lesions was investigated in this study. DHP was administered to male F344 rats via gavage

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service