ZRB3310
Anti-APOBEC3B Antibody, clone 5210-87-133 ZooMAb® Rabbit Monoclonal
recombinant, expressed in HEK 293 cells
Synonym(s):
A3B, Phorbolin-2/3
About This Item
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
5210-87-133, recombinant monoclonal
description
recombinant, expressed in HEK 293 cells
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 46 kDa
observed mol wt ~36 kDa
purified by
using Protein A
species reactivity
human
packaging
antibody small pack of 25
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
technique(s)
ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG
epitope sequence
C-terminal
Protein ID accession no.
UniProt accession no.
greener alternative category
shipped in
ambient
storage temp.
2-8°C
Gene Information
human ... APOBEC3B(9582)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in U2OS cell lysate.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected APOBEC3B in U2OS cell lysate.
Tested Applications
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected APOBEC3B in MCF7 and THP-1 cell lysates.
Immunohistochemistry (Paraffin) Analysis: A 1:25 dilution from a representative lot detected APOBEC3B in Human breast cancer, Human cervical cancer and Human bladder cancer tissue sections.
Enzyme Immunoassay (ELISA) Analysis: A representative lot detected APOBEC3B in ELISA applications (Brown, W., et. al. (2019). Antibodies (Basel). 8(3):47).
Immunocytochemistry Analysis: A 1:25 dilution from a representative lot detected APOBEC3B in U2OS cells.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service