Skip to Content
Merck
All Photos(9)

Documents

ZRB2654

Sigma-Aldrich

Anti-PHACTR1 Antibody, clone 2B15 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Phosphatase and actin regulator 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2B15, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 66.31 kDa
observed mol wt ~60 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

rat, mouse

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

Related Categories

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2B15 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Phosphatase and actin regulator 1 (PHACTR1). It targets an epitope within 23 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 23 amino acids from the N-terminal half of human Phosphatase and actin regulator 1 (PHACTR1).

Application

Quality Control Testing

Evaluated by Western Blotting in U251-MG cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected PHACTR1 in U251-MG cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PHACTR1 in Human brain tissue lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected PHACTR1 in Human cerebral cortex tissue sections.

Immunofluorescence Analysis: A 1:1,000 dilution from a representative lot detected PHACTR1 in Human cerebral cortex tissue sections.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected PHACTR1 in HeLa cells.

Affinity Binding Assay: A representative lot of this antibody bound PHACTR1 peptide with a KD of 1.0 x 10-12in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Phosphatase and actin regulator 1 (UniProt: Q9C0D0; also known as PHACTR1) is encoded by the PHACTR1 (also known as KIAA1733, RPEL1) gene (Gene ID: 221692) in human. PHACTR1 binds actin monomers and plays a role in multiple processes including the regulation of actin cytoskeleton dynamics, actin stress fibers formation, cell motility and survival, formation of tubules by endothelial cells. It contains four RPEL repeats, and it binds three actin monomers via the three C-terminal RPEL repeats. PHACTR1 is also involved in the regulation of cortical neuron migration and dendrite arborization. It is detected in umbilical vein endothelial cells and its levels are up-regulated by vascular endothelial growth factor A (VEGF-A). It resides mainly in cytoplasm in resting cells and is imported into the nucleus upon serum stimulation. Its nuclear localization signal resides in amino acids 108-129. Its expression is up-regulated by pro-inflammatory and pro-atherogenic agents, including TNF- , IL-1β, and oxidized LDL. Mutations in PHACTR1 gene have been linked to developmental and epileptic encephalopathy 70 (DEE70), a heterogeneous group of severe early-onset epilepsies characterized by refractory seizures, neurodevelopmental impairment, and poor prognosis. Polymorphisms in this gene are also associated with susceptibility to myocardial infarction, coronary artery disease and cervical artery dissection. Its interaction with heat shock protein A8 (HSPA8) is reported to be associated with coronary artery disease and eNOS degradation. Two isoforms of PHACTR1 have been described that are produced by alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Rezvan, A. (2023). Arterioscler. Thromb. Vasc. Biol. 43(8); 1409-1411; Ma, X., et al. (2022). Front. Immunol. 13; 958677; Hamada, N., et al. (2018). Brain. 141(11); 3098-3114; Jarray, R., et al. (2011). Biochimie. 93(10); 1668-1675).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service