Skip to Content
Merck
All Photos(12)

Key Documents

ZRB1623

Sigma-Aldrich

Anti-PKCδ Antibody, clone 1C13 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

EC:2.7.11.13, Protein kinase C delta type, nPKC-delta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C13, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 77.51 kDa
observed mol wt ~77 kDa

species reactivity

rat, mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PRKCD(5580)
mouse ... Prkcd(18753)
rat ... Prkcd(170538)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1C13 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects PKCd It targets an epitope within 18 amino acids from the C-terminal, AGC kinase domain.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the C-terminal AGC kinase domain of human protein kinase C delta ( PKCd).

Application

Anti-PKCd, clone 1C13 ZooMAb, Cat. No. ZRB1623, is a recombinant Rabbit monoclonal antibody that specifically targets PKCd and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry, and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Integrin -5 in HT-1080 and NIH3T3 cell lysates.

Flow Cytometry Analysis: 1 μg from a representative lot detected Integrin -5 in one million HelL cells.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected Integrin -5 in human placenta tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound Integrin a-5 peptide with a KD of 2.5 x 10-8 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:1,0000 dilution from a representative lot detected Integrin -5 in HeLa cell line, A431 cell line, HUVEC cell line and NIH 3T3 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Protein kinase Cd (UniProt: Q05655; also known as EC:2.7.11.13, nPKC-delta) is encoded by the PRKCD gene (Gene ID: 5580) in human. PKC d is a ubiquitous serine/threonine kinase involved in signal transduction associated with cell proliferation, differentiation, and apoptosis. It is a calcium-independent, phospholipid- and diacylglycerol (DAG)-dependent novel PKC enzyme that plays contrasting roles in cell death and cell survival by functioning as a pro-apoptotic protein during DNA damage-induced apoptosis but acting as an anti-apoptotic protein during cytokine receptor-initiated cell death. It is shown to negatively regulate B cell proliferation and is also involved in self-antigen induced B cell tolerance induction. Upon DNA damage, PKCd activates the promoter of the death-promoting transcription factor BCLAF1/Btf to trigger BCLAF1-mediated p53/TP53 gene transcription and apoptosis. In the case of ER stress or DNA damage-induced apoptosis, it can form a complex with the tyrosine-protein kinase ABL1 which trigger apoptosis independently of p53/TP53. In cytosol it can trigger apoptosis by activating MAPK11 or MAPK14 and inhibiting Akt1 and decreasing the level of XIAP. Phosphorylation at three specific sites: threonine 507, serine 645, and serine 664 is required for its full activation. PKCd is proteolytically cleaved into a catalytic subunit (aa 330-676) and a regulatory subunit (aa 1-329) by caspase-3 during apoptosis which results in kinase activation. As a result of this cleavage the pseudosubstrate motif in the regulatory subunit is released from the substrate recognition site of the catalytic subunit, which enables its activation. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service