Skip to Content
Merck
All Photos(16)

Key Documents

ZRB1570

Sigma-Aldrich

Anti-Insulin Receptor β Antibody, clone 1J16 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

EC:2.7.10.1;IR;CD220

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
CHF 223.00
4 X 25 μL
CHF 457.00

CHF 223.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
CHF 223.00
4 X 25 μL
CHF 457.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 223.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1J16, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 156.33 kDa
observed mol wt ~110 kDa

purified by

using Protein A

species reactivity

rat, human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1J16 is a Rabbit recombinant monoclonal antibody that specifically detects Insulin receptor. It targets an epitope within 17 amino acids from the C-terminal cytoplasmic domain of the b-subunit.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the C-terminal, cytoplasmic domain of human Insulin receptor b-subunit.

Application

Quality Control Testing

Evaluated by Western Blotting in rat liver tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Insulin Receptor β in rat liver tissue lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected recombinant human Insulin Receptor β and Insulin Receptor b in lysates from Human Kidney and NIH3T3 and U2OS cells.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected Insulin Receptor β in U2OS cells.

Affinity Binding Assay: A representative lot of this antibody bound Insulin Receptor b with a KD of 2.2 x 10-7 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Insulin Receptor β in human kidney tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Insulin receptor (UniProt: P06213; also known as EC:2.7.10.1, IR, CD220) is encoded by the INSR gene (Gene ID: 3643) in human. Insulin is the major hormone that counters the concerted action of a number of hyperglycemia generating and enhances glucose uptake in muscle and adipose tissue, and reduces gluconeogenesis and lipolysis. Its actions are mediated via binding to the Insulin receptor. Insulin receptor is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-27), which is subsequently cleaved off to generate the mature form. It is a tetrameric receptor composed of two a and two β chains linked by disulfide bonds. The a chains carry the insulin-binding region, while the β chains carry the kinase domain. Upon insulin binding, the receptor undergoes autophosphorylation that activates its kinase activity localized in the β subunit. The tyrosine kinase activity then leads to phosphorylation of several intracellular substrates, including, insulin receptor substrates (IRS1, 2, 3, 4), SHC, GAB1, CBL and other signaling intermediates. These phosphorylated proteins serve as docking proteins for other signaling proteins that contain SH2 domains that specifically recognize different phosphotyrosine residues, including the p85 regulatory subunit of PI3K and SHP2. Phosphorylation of IRSs proteins lead to the activation of the PI3K-AKT/PKB pathway, which is responsible for most of the metabolic actions of insulin, and the Ras-MAPK pathway, which regulates expression of some genes and cooperates with the PI3K pathway to control cell growth and differentiation. Upon insulin binding the receptor undergoes autophosphorylation that activates its kinase activity localized in the β subunit. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. ZRB1570 Sigma-Aldrich Anti-Insulin Receptor β Antibody, clone 1J16 ZooMAb® Rabbit Monoclonal What is the 17 amino acid sequence it has been made against?

    1 answer
    1. The exact sequence is proprietary.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service