Skip to Content
Merck
All Photos(11)

Key Documents

ZRB1015

Sigma-Aldrich

Anti-Irisin (FNDC5) Antibody, clone 2K10 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Fibronectin type III domain-containing protein 5, Fibronectin type III repeat-containing protein 2, Irisin

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
CHF 201.00
4 X 25 μL
CHF 415.00

CHF 201.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
CHF 201.00
4 X 25 μL
CHF 415.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 201.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2K10, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 23.66 kDa (calculated for FNDC5. )
observed mol wt ~24 kDa

species reactivity

human

species reactivity (predicted by homology)

rat, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... FNDC5(252995)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 2K10 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Fibronectin type III domain-containing protein 5 (FNDC5). It targets and epitope within the Irisin region.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the Irisin region of the extracellular domain of human Fibronectin type III domain-containing protein 5 (FNDC5).

Application

Anti-Irisin (FNDC5), clone 2K10 ZooMAb, Cat. No. ZRB1015, is a recombinant Rabbit monoclonal antibody that specifically targets Irisin (FNDC5) and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Irisin (FNDC5) in HepG2 cell lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Irisin (FNDC5) in human skeletal muscle cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Irisin (FNDC5) in human skeletal muscle tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound FNDC5 peptide with a KD of 4.5 x 10-9 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Fibronectin type III domain-containing protein 5 (UniProt: Q8NAU1; also known as Fibronectin type III repeat-containing protein 2; FNDC5) is encoded by the FNDC5 (also known as FRCP2) gene (Gene ID: 252995) in human. FNDC5 is a single-pass type I membrane protein that is synthesized with a signal peptide (aa 1-31), which is subsequently cleaved off to produce the mature protein that contains an extracellular domain (aa 32-152), a transmembrane domain (aa 153-173), and a cytoplasmic domain (aa 174-212). The mature FNDC5 can be further cleaved into Irisin (aa 32-143). FNDC5/Irisin is widely expressed and is considered to be a thermogenic, exercise-induced chemokine that is involved in the browning of adipose tissue. It is released mainly by the white subcutaneous adipose tissue and plays a regulatory role in conversion of white fat to brown fat and has a potential in curbing fat accumulation. Its levels are shown to be upregulated by exercise and it can be detected in the blood of individuals following endurance exercise. It has also been associated with reduction of pro-inflammatory cytokines while promoting secretion of anti-inflammatory cytokines in adipose tissue. Irisin is reported to facilitate glucose uptake by skeletal muscles and improve hepatic glucose and lipid metabolism thereby causing a positive effect on hyperlipidemia and hyperglycemia caused by obesity and metabolic syndrome. Lower levels of Irisin are reported in diabetic patients compared to normal subjects. Irisin stimulation of glucose uptake in muscle tissue involves the activation of AMPK pathway. (Ref.: Arhire LI., et al. (2019). Front Endocrinol.10; 524; Zhang, Y., et al. (2014). Diabetes. 63(2); 514-525).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service