Key Documents
Z768642
Nalgene® square graduated media bottle
with cap, PET, sterile, capacity 250 mL
Synonym(s):
media storage bottle, square plastic bottle
Select a Size
60 EA
CHF 306.00
CHF 306.00
Check Cart for Availability
Select a Size
Change View
60 EA
CHF 306.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
CHF 306.00
Check Cart for Availability
material
PET
square bottle
Joint
Joints threaded neck (38-430)
sterility
sterile
feature
graduations
with cap
packaging
pack of 60 ea (2 x, pack of 30)
manufacturer/tradename
Nalgene® 342040-0250
capacity
250 mL
General description
Nalgene® square, graduated bottles are made from polyethylene terephthalate (PET) and are ideal for storing and transporting liquid samples. Convenient, space-saving, light-weight square bottles resist shattering and include a neck ring for tamper-evident and leak-proof seal. High quality resins are non-pyrogenic and non-cytotoxic and come withs a high-density polyethylene (HDPE) closure for sample protection.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)