Z757780
Sartorius filter pipette tips
volume range 2-120 μL, sterile, pack of 960 ea (10 racks of 96 tips)
Synonym(s):
filter pipette tips, pipetting tips, pipettor tips
All Photos(1)
About This Item
Recommended Products
material
colorless
polypropylene (Virgin)
sterility
sterile
feature
barrier
filter
packaging
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Sartorius 790101F
volume range
2-120 μL
binding type
low binding surface
Related Categories
General description
The Sartorius filter tips of the Biohitfamily are made of polypropylene and feature a polyethylene filter barrier to prevent aerosol and liquid contamination. This filter helps protect against the rist of cross-contamination and reduces maintenance requirements of the pipette. The filter tips are suited for molecular biology, microbiology, cell culture and radioactive work.
These tips have more space between the sample and the filter than conventional filter tips so you do not need to worry about the sample touching the filter whatever the pipetting technique or type of liquid.
Benefits:
These tips have more space between the sample and the filter than conventional filter tips so you do not need to worry about the sample touching the filter whatever the pipetting technique or type of liquid.
Benefits:
- Pipette foaming liquids like buffers and proteins
- Pipette solvents
- Reverse pipetting
- Multiple dispensing functions of electronic pipettes
Other Notes
Browse all pipette tips