Skip to Content
Merck
All Photos(2)

Key Documents

Z757462

Sartorius Proline® Plus multichannel mechanical pippettor

8-channel, volume 30-300 μL

Synonym(s):

Multichannel Pipettor

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CHF 860.00

CHF 860.00


Check Cart for Availability


Select a Size

Change View
1 EA
CHF 860.00

About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

CHF 860.00


Check Cart for Availability

feature

8-channel
electronic: no

packaging

pack of 1 ea

manufacturer/tradename

Sartorius 728140

volume

30-300 μL

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • The click-stop system enables easy volume adjustment.
  • Broad volume display for better reading.
  • Color-coded for easy matching of fitting tips
  • High chemical and UV-resistance materials to ensure a long lifetime of the pipette.
  • Fully autoclavable and requires no disassembly.
  • Easy cleaning and maintenance with only three parts to disassemble.
  • Safe-cone filters provided for models >10μL.
  • Εrgοnomic finger support reduces the grip force required to hold the pipette.
  • Range of volume-3μL (5μL for fixed) to 10mL
  • Low pipetting forces
  • Wide range of adjustable single and multi-channel models and fixed single-channel models

Legal Information

Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service