Key Documents
Z757330
Sartorius Proline® Plus mechanical pippettor
single channel, volume 0.1-3 μL
Select a Size
1 EA
CHF 293.00
CHF 293.00
Estimated to ship on24 April 2025
Select a Size
Change View
1 EA
CHF 293.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
CHF 293.00
Estimated to ship on24 April 2025
feature
electronic: no
single-channel
packaging
pack of 1 ea
manufacturer/tradename
Sartorius 728010
volume
0.1-3 μL
General description
The Sartorius Proline Plus pipettes of the Biohitfamily are high quality with a light pipetting force. With robust design, the Proline Plus is suitable for variable every day laboratory applications and covers the full range of 0.1μL to 10 mL.
  • Reduced pipetting forces prevent RSI and improve the results
  • Accurate and precise
  • Contamination free pipetting with Safe-Cone Filters
  • Robust design
  • Easiest pipette to maintain
Legal Information
Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy
Choose from one of the most recent versions:
Certificates of Analysis (COA)